Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4-Dimethyl-1,2,3,4-tetrahydroquinoline
4,4-Dimethyl-1,2,3,4-tetrahydroquinoline 96%
4,4-Dimethyl-1,2,3,4-tetrahydroquinoline 96%
4,4-Dimethyl-1,2,3,4-tetrahydroquinoline 96%
Supplier:
Apollo Scientific
Certificates
Expand image
4,4-Dimethyl-1,2,3,4-tetrahydroquinoline 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
20364-31-2
:
Formula:
C₁₁H₁₅N
MDL Number:
MFCD11847806
:
161.243 g/mol
Storage Temperature:
Ambient