Order Entry
Export
ContactUsLinkComponent
2,5-Dimethyl-4,6-dihydroxypyrimidine 95%
2,5-Dimethyl-4,6-dihydroxypyrimidine 95%
Catalog # APOSOR918718-1G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dimethyl-4,6-dihydroxypyrimidine 95%
Catalog # APOSOR918718-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :140.14 g/mol
Storage Temperature:Ambient
CAS Number:1194-74-7
Formula:C₆H₈N₂O₂
MDL Number:MFCD02685061