Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methoxy-2,2-dimethylpropan-1-ol
3-Methoxy-2,2-dimethylpropan-1-ol 95%
3-Methoxy-2,2-dimethylpropan-1-ol 95%
APOSOR916644-1G
:
Apollo Scientific
:
3-Methoxy-2,2-dimethylpropan-1-ol 95%
APOSOR916644-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
118.174 g/mol
:
Ambient
:
57021-67-7
:
C₆H₁₄O₂
:
MFCD09908260