Order Entry
Export
ContactUsLinkComponent
 
Methyl 4-methoxy-2,5-dimethylbenzoate 95%
Catalog # APOSOR916559-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-methoxy-2,5-dimethylbenzoate 95%
Catalog # APOSOR916559-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.227 g/mol
Storage Temperature:Ambient
CAS Number:58106-28-8
Formula:C₁₁H₁₄O₃
MDL Number:MFCD06203806