Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-2-Benzyl-aziridine
(S)-2-Benzyl-aziridine 95%
(S)-2-Benzyl-aziridine 95%
(S)-2-Benzyl-aziridine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
(S)-2-Benzyl-aziridine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
73058-30-7
:
Formula:
C₉H₁₁N
MDL Number:
MFCD06796591
:
133.19 g/mol
Storage Temperature:
Ambient