Order Entry
Export
ContactUsLinkComponent
1-Acetyl-5-chloro-pyrazolo-[3,4-c]pyridine 95%
1-Acetyl-5-chloro-pyrazolo-[3,4-c]pyridine 95%
Catalog # APOSOR913288-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Acetyl-5-chloro-pyrazolo-[3,4-c]pyridine 95%
Catalog # APOSOR913288-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.606 g/mol
Storage Temperature:Ambient
CAS Number:76006-04-7
Formula:C₈H₆ClN₃O
MDL Number:MFCD08061149