Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dimethyl-5-nitroaniline
2,3-Dimethyl-5-nitroaniline 95%
2,3-Dimethyl-5-nitroaniline 95%
2,3-Dimethyl-5-nitroaniline 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Dimethyl-5-nitroaniline 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
109508-62-5
:
106837-44-9
Formula:
C₈H₁₀N₂O₂
MDL Number:
MFCD05664520
:
166.177 g/mol
Storage Temperature:
Ambient