Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Aminomethyl)-2,5-dimethyl-1,3-oxazole
4-(Aminomethyl)-2,5-dimethyl-1,3-oxazole
4-(Aminomethyl)-2,5-dimethyl-1,3-oxazole
APOSOR9127-500MG
:
Apollo Scientific
:
4-(Aminomethyl)-2,5-dimethyl-1,3-oxazole
APOSOR9127-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
36…38 °C (0.1 mm Hg)
:
Ambient
:
2735
:
8,III
:
126.156 g/mol
:
GHS05
:
859850-62-7
:
C₆H₁₀N₂O
:
MFCD08271883