Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,4,5-Tetrahydro-1H-pyrido[4,3-b]indole
2,3,4,5-Tetrahydro-1H-pyrido[4,3-b]indole 96+%
2,3,4,5-Tetrahydro-1H-pyrido[4,3-b]indole 96+%
2,3,4,5-Tetrahydro-1H-pyrido[4,3-b]indole 96+%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3,4,5-Tetrahydro-1H-pyrido[4,3-b]indole 96+%
Supplier:
Apollo Scientific
About this item
CAS Number:
6208-60-2
:
Formula:
C₁₁H₁₂N₂
MDL Number:
MFCD00464648
:
172.226 g/mol
Storage Temperature:
Ambient