Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Butyl-pyrrole-2,5-dione
1-Butyl-pyrrole-2,5-dione 95%
1-Butyl-pyrrole-2,5-dione 95%
APOSOR912213-1G
:
Apollo Scientific
:
1-Butyl-pyrrole-2,5-dione 95%
APOSOR912213-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
153.178 g/mol
:
Ambient
:
2973-09-3
:
C₈H₁₁NO₂
:
MFCD00042756