Order Entry
Export
ContactUsLinkComponent
3-(3,5-Dimethylphenyl)aniline 95%
3-(3,5-Dimethylphenyl)aniline 95%
Catalog # APOSOR910878-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(3,5-Dimethylphenyl)aniline 95%
Catalog # APOSOR910878-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.276 g/mol
Storage Temperature:Ambient
CAS Number:783325-73-5
Formula:C₁₄H₁₅N
MDL Number:MFCD12779833