5-Methoxy-1-methyl-1,3-benzodiazole 96%
Catalog # APOSOR910453-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.188 g/mol
Storage Temperature:Ambient
CAS Number:10394-39-5
Formula:C₉H₁₀N₂O
MDL Number:MFCD00962665