Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(2,5-Dimethylpyrazol-3-yl)acetamide
N-(2,5-Dimethylpyrazol-3-yl)acetamide 96%
N-(2,5-Dimethylpyrazol-3-yl)acetamide 96%
N-(2,5-Dimethylpyrazol-3-yl)acetamide 96%
Supplier:
Apollo Scientific
Certificates
Expand image
N-(2,5-Dimethylpyrazol-3-yl)acetamide 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
75092-37-4
:
Formula:
C₇H₁₁N₃O
MDL Number:
MFCD01566426
:
153.182 g/mol
Storage Temperature:
Ambient