0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-Methoxy-1,3-benzothiazole
4-Methoxy-1,3-benzothiazole 95%
4-Methoxy-1,3-benzothiazole 95%
4-Methoxy-1,3-benzothiazole 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Methoxy-1,3-benzothiazole 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
3048-46-2
:
Formula:
C₈H₇NOS
MDL Number:
MFCD16877170
:
165.212 g/mol
Storage Temperature:
Ambient