Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Cyclopropyl-2,5-dimethylpyrrole
1-Cyclopropyl-2,5-dimethylpyrrole 95%
1-Cyclopropyl-2,5-dimethylpyrrole 95%
1-Cyclopropyl-2,5-dimethylpyrrole 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-Cyclopropyl-2,5-dimethylpyrrole 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
54609-12-0
:
Formula:
C₉H₁₃N
MDL Number:
MFCD12187123
:
135.206 g/mol
Storage Temperature:
Ambient