4-Bromo-1,5-dimethylimidazole 95%
Catalog # APOSOR910018-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:175.026 g/mol
Storage Temperature:Ambient
CAS Number:158585-81-0
Formula:C₅H₇BrN₂
MDL Number:MFCD09033089