Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dimethyl-1,3-benzothiazol-2-amine
4,5-Dimethyl-1,3-benzothiazol-2-amine 98%
4,5-Dimethyl-1,3-benzothiazol-2-amine 98%
4,5-Dimethyl-1,3-benzothiazol-2-amine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
4,5-Dimethyl-1,3-benzothiazol-2-amine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
79050-49-0
:
Formula:
C₉H₁₀N₂S
MDL Number:
MFCD03764730
:
178.254 g/mol
Storage Temperature:
Ambient