Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(3-Isocyanopropyl)-n,n-dimethylamine
N-(3-Isocyanopropyl)-n,n-dimethylamine 95%
N-(3-Isocyanopropyl)-n,n-dimethylamine 95%
N-(3-Isocyanopropyl)-n,n-dimethylamine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
N-(3-Isocyanopropyl)-n,n-dimethylamine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
15586-24-0
:
Formula:
C₆H₁₂N₂
MDL Number:
MFCD02664631
:
112.173 g/mol
Storage Temperature:
Ambient