(5,6-Dimethoxypyridin-3-yl)boronic acid 95%
Catalog # APOSOR906990-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.97 g/mol
Storage Temperature:Ambient
CAS Number:1346526-61-1
Formula:C₇H₁₀BNO₄
MDL Number:MFCD18254414