Order Entry
Export
ContactUsLinkComponent
 
4-(4-Methylphenyl)-1,3-thiazole 95%
Catalog # APOSOR905477-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(4-Methylphenyl)-1,3-thiazole 95%
Catalog # APOSOR905477-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.25 g/mol
Storage Temperature:Ambient
CAS Number:1826-19-3
Formula:C₁₀H₉NS
MDL Number:MFCD02665781