Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2,4-Dimethylphenyl)pyrrolidine
2-(2,4-Dimethylphenyl)pyrrolidine 95%
2-(2,4-Dimethylphenyl)pyrrolidine 95%
APOSOR905265-1G
:
Apollo Scientific
:
2-(2,4-Dimethylphenyl)pyrrolidine 95%
APOSOR905265-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
175.27 g/mol
:
Ambient
:
298690-87-6
:
C₁₂H₁₇N
:
MFCD02663504