Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2,4-Dimethylphenyl)piperidine
2-(2,4-Dimethylphenyl)piperidine 95%
2-(2,4-Dimethylphenyl)piperidine 95%
2-(2,4-Dimethylphenyl)piperidine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2-(2,4-Dimethylphenyl)piperidine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
383128-60-7
:
Formula:
C₁₃H₁₉N
MDL Number:
MFCD02663651
:
189.297 g/mol
Storage Temperature:
Ambient