
2-(2,4-Dichlorophenyl)imidazo[1,2-a]pyridine
Catalog # APOSOR904619-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:263.122 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:63111-80-8
Formula:C₁₃H₈Cl₂N₂
MDL Number:MFCD01191735