Order Entry
Export
ContactUsLinkComponent
1-Methyl-2,2'-bipiperidine 97%
1-Methyl-2,2'-bipiperidine 97%
Catalog # APOSOR904408-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-2,2'-bipiperidine 97%
Catalog # APOSOR904408-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.306 g/mol
Storage Temperature:Ambient
CAS Number:118046-22-3
Formula:C₁₁H₂₂N₂
MDL Number:MFCD02663572