Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyrrolo[2,1-f][1,2,4]triazine-2,4(1H,3H)-dione
Pyrrolo[2,1-f][1,2,4]triazine-2,4(1H,3H)-dione 95%
Pyrrolo[2,1-f][1,2,4]triazine-2,4(1H,3H)-dione 95%
Pyrrolo[2,1-f][1,2,4]triazine-2,4(1H,3H)-dione 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Pyrrolo[2,1-f][1,2,4]triazine-2,4(1H,3H)-dione 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
918538-04-2
:
Formula:
C₆H₅N₃O₂
MDL Number:
MFCD11053713
:
151.123 g/mol
Storage Temperature:
Ambient