Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Isopropyl-5-methylimidazolidine-2,4-dione
5-Isopropyl-5-methylimidazolidine-2,4-dione 96%
5-Isopropyl-5-methylimidazolidine-2,4-dione 96%
5-Isopropyl-5-methylimidazolidine-2,4-dione 96%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Isopropyl-5-methylimidazolidine-2,4-dione 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
150226-22-5
:
Formula:
C₇H₁₂N₂O₂
MDL Number:
MFCD00033554
:
156.182 g/mol
Storage Temperature:
Ambient