Order Entry
Export
ContactUsLinkComponent
1,3-Dicyclopentylurea 95%
1,3-Dicyclopentylurea 95%
Catalog # APOSOR900409-1G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Dicyclopentylurea 95%
Catalog # APOSOR900409-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :196.289 g/mol
Storage Temperature:Ambient
CAS Number:58713-33-0
Formula:C₁₁H₂₀N₂O
MDL Number:MFCD13258925