Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2-Hydroxyphenyl)pyrrolidine-2,5-dione
1-(2-Hydroxyphenyl)pyrrolidine-2,5-dione 98%
1-(2-Hydroxyphenyl)pyrrolidine-2,5-dione 98%
1-(2-Hydroxyphenyl)pyrrolidine-2,5-dione 98%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(2-Hydroxyphenyl)pyrrolidine-2,5-dione 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
79252-84-9
:
Formula:
C₁₀H₉NO₃
MDL Number:
MFCD00461771
:
191.183 g/mol
Storage Temperature:
Ambient