Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2,5-dibromopentanoate
Methyl 2,5-dibromopentanoate
Methyl 2,5-dibromopentanoate
Irritant
Methyl 2,5-dibromopentanoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 2,5-dibromopentanoate
Supplier:
Apollo Scientific
About this item
CAS Number:
50995-48-7
:
Formula:
C₆H₁₀Br₂O₂
MDL Number:
MFCD02684289
:
273.95 g/mol
:
GHS07
Storage Temperature:
Ambient