0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,7,7-Trimethylbicyclo[2.2.1]heptane
1,7,7-Trimethylbicyclo[2.2.1]heptane 95+%
1,7,7-Trimethylbicyclo[2.2.1]heptane 95+%
APOSOR7980-250MG
:
Apollo Scientific
:
1,7,7-Trimethylbicyclo[2.2.1]heptane 95+%
APOSOR7980-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
138.25 g/mol
:
156…159 °C
:
GHS07
:
Ambient
:
464-15-3
:
C₁₀H₁₈
:
MFCD00213517