
1H-Indole-2-carbohydrazide
Catalog # APOSOR7963-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:175.187 g/mol
Melting Pt:243…245 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:5055-39-0
Formula:C₉H₉N₃O
MDL Number:MFCD02293210