Order Entry
Export
ContactUsLinkComponent
 
Irritant
2-(1,6-Naphthyridin-2-yl)benzoic acid hydrate 95+%
Catalog # APOSOR7867-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(1,6-Naphthyridin-2-yl)benzoic acid hydrate 95+%
Catalog # APOSOR7867-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :268.267 g/mol
Melting Pt:175…185 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:1172902-78-1
Formula:C₁₅H₁₀N₂O₂·H₂O
MDL Number:MFCD08445563