
4-[(Benzyloxy)methyl]-1,3-dioxolan-2-one
Catalog # APOSOR7801T-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:208.211 g/mol
Boiling Pt:165 °C (0.8 mm Hg)
:GHS07
Storage Temperature:Refrigerator
CAS Number:949-97-3
Formula:C₁₁H₁₂O₄
MDL Number:MFCD00269930