Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Furan-2-yl)benzaldehyde
4-(Furan-2-yl)benzaldehyde
4-(Furan-2-yl)benzaldehyde
4-(Furan-2-yl)benzaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
4-(Furan-2-yl)benzaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
60456-77-1
:
Formula:
C₁₁H₈O₂
MDL Number:
MFCD03030050
:
172.18 g/mol
Storage Temperature:
Ambient