Order Entry
Export
ContactUsLinkComponent
4'-(Benzyloxy)-[1,1'-biphenyl]-2-amine 98%
Irritant
4'-(Benzyloxy)-[1,1'-biphenyl]-2-amine 98%
Catalog # APOSOR7323-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4'-(Benzyloxy)-[1,1'-biphenyl]-2-amine 98%
Catalog # APOSOR7323-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :275.344 g/mol
Melting Pt:116…118 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:400746-75-0
Formula:C₁₉H₁₇NO
MDL Number:MFCD04039047