
3'-Amino-[1,1'-biphenyl]-3-carboxylic acid hydrochloride
Catalog # APOSOR7317-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:249.693 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1170469-44-9
Formula:C₁₃H₁₁NO₂·ClH
MDL Number:MFCD08456987