
Camphorquinone-10-sulphonic acid hydrate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:73413-79-3
:
Formula:C₁₀H₁₄O₅S·H₂O
MDL Number:MFCD00269942
:264.295 g/mol
Melting Pt:138…140 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05