Order Entry
Export
ContactUsLinkComponent
4,6-Dichloropyrimidine-2-carboxylic acid 95%
Irritant
4,6-Dichloropyrimidine-2-carboxylic acid 95%
Catalog # APOSOR70201-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,6-Dichloropyrimidine-2-carboxylic acid 95%
Catalog # APOSOR70201-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.988 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:684220-30-2
Formula:C₅H₂Cl₂N₂O₂
MDL Number:MFCD09834770