Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4'-Dimethylbiphenyl
2,4'-Dimethylbiphenyl
2,4'-Dimethylbiphenyl
Irritant
2,4'-Dimethylbiphenyl
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4'-Dimethylbiphenyl
Supplier:
Apollo Scientific
About this item
CAS Number:
611-61-0
:
Formula:
C₁₄H₁₄
MDL Number:
MFCD15042185
:
182.261 g/mol
Boiling Pt:
273…276 °C
Density:
0.9924 g/cm³ (25 °C)
:
GHS07
Storage Temperature:
Ambient