Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4'-Dimethylbiphenyl
3,4'-Dimethylbiphenyl
3,4'-Dimethylbiphenyl
Irritant
3,4'-Dimethylbiphenyl
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,4'-Dimethylbiphenyl
Supplier:
Apollo Scientific
About this item
CAS Number:
7383-90-6
:
Formula:
C₁₄H₁₄
MDL Number:
MFCD00602700
:
182.261 g/mol
:
GHS07
Storage Temperature:
Ambient