

4-Methyl-1,3-oxazole-5-carbonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1003-52-7
:
Formula:C₅H₄N₂O
MDL Number:MFCD01111976
:108.098 g/mol
Boiling Pt:162 °C (763 mm Hg)
Density:1.12 g/cm³ (25 °C)
Flash Pt:60 °C
Storage Temperature:Ambient
EINECS:213-709-7
UN:1993
ADR:3,III
:GHS02
:GHS07