
1-(1,3-Thiazol-2-yl)thiourea
Catalog # APOSOR6809-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:159.233 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:19958-82-8
Formula:C₄H₅N₃S₂
MDL Number:MFCD04971712