Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,6-Anhydro-β-D-mannopyranose
1,6-Anhydro-β-D-mannopyranose
1,6-Anhydro-β-D-mannopyranose
1,6-Anhydro-β-D-mannopyranose
Supplier:
Apollo Scientific
Certificates
Expand image
1,6-Anhydro-β-D-mannopyranose
Supplier:
Apollo Scientific
About this item
CAS Number:
14168-65-1
:
Formula:
C₆H₁₀O₅
MDL Number:
MFCD00069698
:
162.141 g/mol
Melting Pt:
185 °C
:
GHS07
Storage Temperature:
Ambient