15-Phenylpentadecanoic acid
Catalog # APOSOR6471-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:318.493 g/mol
Melting Pt:59…61 °C
:GHS07
Storage Temperature:Ambient
CAS Number:40228-93-1
Formula:C₂₁H₃₄O₂
MDL Number:MFCD00040777