Order Entry
Export
ContactUsLinkComponent
 
5-Ethynyl-1H-pyrazolo[4,3-b]pyridine
Catalog # APOSOR62078-250MG
Supplier:  Apollo Scientific
CAS Number:  
5-Ethynyl-1H-pyrazolo[4,3-b]pyridine
Catalog # APOSOR62078-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :143.145 g/mol
Storage Temperature:Ambient
CAS Number:1374652-56-8
Formula:C₈H₅N₃
MDL Number:MFCD22380512