Order Entry
Export
ContactUsLinkComponent
2-Bromo-5,5-dimethyl-5,6-dihydrobenzo[d]thiazol-7(4H)-one 98%
Irritant
2-Bromo-5,5-dimethyl-5,6-dihydrobenzo[d]thiazol-7(4H)-one 98%
Catalog # APOSOR61089-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Bromo-5,5-dimethyl-5,6-dihydrobenzo[d]thiazol-7(4H)-one 98%
Catalog # APOSOR61089-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :260.151 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:10513-26-5
Formula:C₉H₁₀BrNOS
MDL Number:MFCD20527744