
4-(Methylsulphonyl)benzene-1,2-diamine
Catalog # APOSOR61025-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:186.232 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:21731-57-7
Formula:C₇H₁₀N₂O₂S
MDL Number:MFCD00720917