Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Imidazo[1,2-a]pyridine-6-carboxamide
Imidazo[1,2-a]pyridine-6-carboxamide
Imidazo[1,2-a]pyridine-6-carboxamide
Irritant
Imidazo[1,2-a]pyridine-6-carboxamide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Imidazo[1,2-a]pyridine-6-carboxamide
Supplier:
Apollo Scientific
About this item
CAS Number:
103313-38-8
:
Formula:
C₈H₇N₃O
MDL Number:
MFCD13176747
:
161.161 g/mol
Melting Pt:
202…204 °C
:
GHS07
Storage Temperature:
Ambient