
5-(Hydroxymethyl)-2-methyl-1,3-oxazole 95%
Catalog # APOSOR59863-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:113.115 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1065073-48-4
Formula:C₅H₇NO₂
MDL Number:MFCD11101415