0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-(Hydroxymethyl)-2-methyl-1,3-oxazole
4-(Hydroxymethyl)-2-methyl-1,3-oxazole 98%
4-(Hydroxymethyl)-2-methyl-1,3-oxazole 98%
Irritant
4-(Hydroxymethyl)-2-methyl-1,3-oxazole 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-(Hydroxymethyl)-2-methyl-1,3-oxazole 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
141567-53-5
:
Formula:
C₅H₇NO₂
MDL Number:
MFCD08703638
:
113.115 g/mol
:
GHS07
Storage Temperature:
Refrigerator